Assay Details
| Gene Symbol: YWHAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284545.1 | NP_001271474.1 | 1 - 2 | 236 | 80 | |
| XM_015437385.1 | XP_015292871.1 | 1 - 2 | 236 | 80 | ||
| XM_015437386.1 | XP_015292872.1 | 1 - 2 | 197 | 80 | ||
| GenBank mRNA | AB169901.1 | - | 1 - 2 | 158 | 80 | |
| AB171120.1 | - | 1 - 2 | 236 | 80 | ||
More Information
Set Membership: |
|

YWHAE
Genomic Map