Assay Details
| Gene Symbol: MIF4GD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584913.1 | XP_005584970.1 | 3 - 4 | 520 | 64 | |
| XM_005584914.2 | XP_005584971.1 | 3 - 4 | 286 | 64 | ||
More Information
Set Membership: |
|

MIF4GD
Genomic Map