Assay Details
| Gene Symbol: NIP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283813.1 | NP_001270742.1 | 2 - 3 | 149 | 150 | |
| GenBank mRNA | AB173512.1 | - | 2 - 3 | 149 | 150 | |
More Information
Set Membership: |
|

NIP7
Genomic Map