Assay Details
| Gene Symbol: SH3BGRL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544340.2 | XP_005544397.1 | 1 - 2 | 999 | 89 | |
| GenBank mRNA | AB171552.1 | - | 1 - 2 | 104 | 89 | |
More Information
Set Membership: |
|

SH3BGRL3
Genomic Map