Assay Details
| Gene Symbol: SH2D2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541401.2 | XP_005541458.1 | 3 - 4 | 459 | 83 | |
| XM_005541402.2 | XP_005541459.1 | 3 - 4 | 447 | 83 | ||
| XM_005541403.2 | XP_005541460.1 | 3 - 4 | 429 | 83 | ||
| XM_015455478.1 | XP_015310964.1 | 3 - 4 | 417 | 83 | ||
| XM_015455482.1 | XP_015310968.1 | 3 - 4 | 462 | 83 | ||
More Information
Set Membership: |
|

SH2D2A
Genomic Map