Assay Details
| Gene Symbol: EAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005545724.2 | XP_005545781.1 | 3 - 4 | 1141 | 67 | |
| GenBank mRNA | AB168382.1 | - | 3 - 4 | 293 | 67 | |
More Information
Set Membership: |
|

EAF1
Genomic Map