Assay Details
| Gene Symbol: HTR1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544486.2 | XP_005544543.1 | 3 | 1809 | 55 | |
| XM_015441004.1 | XP_015296490.1 | 4 | 1868 | 55 | ||
| XM_015441015.1 | XP_015296501.1 | 2 | 1753 | 55 | ||
More Information
Set Membership: |
|

HTR1D
Genomic Map