Assay Details
| Gene Symbol: ZNF445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546846.2 | XP_005546903.1 | 5 - 6 | 1084 | 76 | |
| XM_005546847.2 | XP_005546904.1 | 5 - 6 | 1048 | 76 | ||
| XM_005546848.2 | XP_005546905.1 | 3 - 4 | 337 | 76 | ||
| XM_015445744.1 | XP_015301230.1 | 6 - 7 | 1139 | 76 | ||
More Information
Set Membership: |
|

ZNF445
Genomic Map