Assay Details
| Gene Symbol: ARHGAP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565169.2 | XP_005565226.1 | 6 - 7 | 1194 | 66 | |
| XM_005565171.2 | XP_005565228.1 | 7 - 8 | 1733 | 66 | ||
| XM_005565172.2 | XP_005565229.1 | 6 - 7 | 1146 | 66 | ||
| XM_005565173.2 | XP_005565230.2 | 4 - 5 | 7114 | 66 | ||
| XM_015456054.1 | XP_015311540.1 | 7 - 8 | 1781 | 66 | ||
More Information
Set Membership: |
|

ARHGAP22
Genomic Map