Assay Details
| Gene Symbol: MAML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579421.2 | XP_005579478.1 | 5 - 6 | 2904 | 76 | |
| XM_005579422.2 | XP_005579479.1 | 4 - 5 | 14982 | 76 | ||
| XM_015435625.1 | XP_015291111.1 | 4 - 5 | 3861 | 76 | ||
| XM_015435626.1 | XP_015291112.1 | 4 - 5 | 2267 | 76 | ||
More Information
Set Membership: |
|

MAML2
Genomic Map