Assay Details
| Gene Symbol: MBD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015432173.1 | XP_015287659.1 | 6 - 7 | 7628 | 91 | |
| XM_015432174.1 | XP_015287660.1 | 7 - 8 | 3392 | 91 | ||
| XM_015432175.1 | XP_015287661.1 | 8 - 9 | 6251 | 91 | ||
| XM_015432176.1 | XP_015287662.1 | 5 - 6 | 3710 | 91 | ||
| XM_015432177.1 | XP_015287663.1 | 4 - 5 | 6342 | 91 | ||
| XM_015432178.1 | XP_015287664.1 | 5 - 6 | 3854 | 91 | ||
| XM_015432179.1 | XP_015287665.1 | 7 - 8 | 7386 | 91 | ||
| XM_015432180.1 | XP_015287666.1 | 7 - 8 | 7775 | 91 | ||
| XM_015432181.1 | XP_015287667.1 | 7 - 8 | 7775 | 91 | ||
| XM_015432182.1 | XP_015287668.1 | 7 - 8 | 7775 | 91 | ||
| XM_015432183.1 | XP_015287669.1 | 7 - 8 | 7775 | 91 | ||
| XM_015432184.1 | XP_015287670.1 | 8 - 9 | 4159 | 91 | ||
| XM_015432185.1 | XP_015287671.1 | 9 - 10 | 4294 | 91 | ||
| XM_015432186.1 | XP_015287672.1 | 8 - 9 | 5073 | 91 | ||
More Information
Set Membership: |
|

MBD5
Genomic Map