Assay Details
| Gene Symbol: THRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285306.1 | NP_001272235.1 | 6 - 7 | 777 | 80 | |
| XM_015444467.1 | XP_015299953.1 | 7 - 8 | 1176 | 80 | ||
| XM_015444472.1 | XP_015299958.1 | 7 - 8 | 1068 | 80 | ||
| XM_015444475.1 | XP_015299961.1 | 7 - 8 | 2366 | 80 | ||
| XM_015444478.1 | XP_015299964.1 | 7 - 8 | 1089 | 80 | ||
| XM_015444483.1 | XP_015299969.1 | 7 - 8 | 939 | 80 | ||
| XM_015444486.1 | XP_015299972.1 | 7 - 8 | 1133 | 80 | ||
| XM_015444492.1 | XP_015299978.1 | 7 - 8 | 1355 | 80 | ||
| XM_015444495.1 | XP_015299981.1 | 3 - 4 | 3247 | 80 | ||
| XM_015444497.1 | XP_015299983.1 | 7 - 8 | 1095 | 80 | ||
| XM_015444499.1 | XP_015299985.1 | 8 - 9 | 1317 | 80 | ||
| XM_015444502.1 | XP_015299988.1 | 7 - 8 | 1476 | 80 | ||
| XM_015444504.1 | XP_015299990.1 | 6 - 7 | 966 | 80 | ||
| XM_015444508.1 | XP_015299994.1 | 8 - 9 | 1233 | 80 | ||
| XM_015444512.1 | XP_015299998.1 | 6 - 7 | 858 | 80 | ||
| XM_015444516.1 | XP_015300002.1 | 6 - 7 | 796 | 80 | ||
| XM_015444520.1 | XP_015300006.1 | 7 - 8 | 1188 | 80 | ||
| XM_015444526.1 | XP_015300012.1 | 5 - 6 | 900 | 80 | ||
| GenBank mRNA | AB172636.1 | - | 6 - 7 | 777 | 80 | |
More Information
Set Membership: |
|

THRB
Genomic Map