Assay Details
| Gene Symbol: MTREX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556905.2 | XP_005556962.1 | 23 - 24 | 3013 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MTREX| Gene Symbol: MTREX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556905.2 | XP_005556962.1 | 23 - 24 | 3013 | 93 | |
Set Membership: |
|