Assay Details
| Gene Symbol: CDK2AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577057.2 | XP_005577114.1 | 1 - 2 | 413 | 101 | |
| XM_005577058.1 | XP_005577115.1 | 2 - 3 | 827 | 101 | ||
| XM_005577060.2 | XP_005577117.1 | 2 - 3 | 426 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005577060.2 transcripts.
Set Membership: |
|

CDK2AP2
Genomic Map