Assay Details
Gene Symbol: PAQR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283351.1 | NP_001270280.1 | 1 - 2 | 176 | 91 | |
XM_005554969.2 | XP_005555026.1 | 1 - 2 | 337 | 91 | ||
XM_005554970.2 | XP_005555027.1 | 1 - 2 | 337 | 91 | ||
XM_005554972.2 | XP_005555029.1 | 1 - 2 | 337 | 91 | ||
XM_005554973.2 | XP_005555030.1 | 1 - 2 | 337 | 91 | ||
XM_015450071.1 | XP_015305557.1 | 1 - 2 | 337 | 91 | ||
XM_015450072.1 | XP_015305558.1 | 1 - 2 | 337 | 91 | ||
XM_015450073.1 | XP_015305559.1 | 1 - 2 | 337 | 91 | ||
GenBank mRNA | AB169058.1 | - | 1 - 2 | 176 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
