Assay Details
| Gene Symbol: LOC102142811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566168.2 | XP_005566225.1 | 3 - 4 | 699 | 109 | |
| XM_005566169.2 | XP_005566226.1 | 3 - 4 | 699 | 109 | ||
| XM_005566170.2 | XP_005566227.1 | 3 - 4 | 699 | 109 | ||
| XM_015456530.1 | XP_015312016.1 | 3 - 4 | 699 | 109 | ||
| GenBank mRNA | AB174381.1 | - | 3 - 4 | 480 | 109 | |
More Information
Set Membership: |
|

LOC102142811
Genomic Map