Assay Details
| Gene Symbol: ATF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570845.2 | XP_005570902.1 | 2 - 3 | 234 | 69 | |
| XM_005570846.2 | XP_005570903.1 | 2 - 3 | 343 | 69 | ||
| XM_005570847.2 | XP_005570904.1 | 2 - 3 | 770 | 69 | ||
| XM_005570848.2 | XP_005570905.1 | 2 - 3 | 826 | 69 | ||
| XM_005570849.2 | XP_005570906.1 | 2 - 3 | 766 | 69 | ||
| XM_015430917.1 | XP_015286403.1 | 3 - 4 | 847 | 69 | ||
| XM_015430919.1 | XP_015286405.1 | 3 - 4 | 414 | 69 | ||
More Information
Set Membership: |
|

ATF1
Genomic Map