Assay Details
| Gene Symbol: CAPZA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285329.1 | NP_001272258.1 | 3 - 4 | 239 | 117 | |
| GenBank mRNA | AB169582.1 | - | 3 - 4 | 239 | 117 | |
More Information
Set Membership: |
|

CAPZA2
Genomic Map