Assay Details
| Gene Symbol: IFRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284759.1 | NP_001271688.1 | 4 - 5 | 660 | 85 | |
| GenBank mRNA | AB169259.1 | - | 4 - 5 | 660 | 85 | |
More Information
Set Membership: |
|

IFRD1
Genomic Map