Assay Details
| Gene Symbol: SEPTIN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591155.2 | XP_005591212.1 | 4 - 5 | 682 | 103 | |
| XM_015442371.1 | XP_015297857.1 | 5 - 6 | 646 | 103 | ||
| XM_015442372.1 | XP_015297858.1 | 4 - 5 | 601 | 103 | ||
More Information
Set Membership: |
|

SEPTIN12
Genomic Map