Assay Details
| Gene Symbol: PRSS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553785.1 | XP_005553842.1 | 8 - 9 | 1042 | 86 | |
| XM_015449754.1 | XP_015305240.1 | 8 - 9 | 1043 | 86 | ||
More Information
Set Membership: |
|

PRSS16
Genomic Map