Assay Details
| Gene Symbol: TBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284983.1 | NP_001271912.1 | 5 - 6 | 908 | 112 | |
| XM_005551358.2 | XP_005551415.1 | 5 - 6 | 1150 | 112 | ||
| XM_005551360.2 | XP_005551417.1 | 5 - 6 | 1052 | 112 | ||
| XM_005551361.2 | XP_005551418.1 | 5 - 6 | 913 | 112 | ||
| XM_005551362.2 | XP_005551419.1 | 6 - 7 | 1072 | 112 | ||
| XM_005551363.2 | XP_005551420.1 | 5 - 6 | 885 | 112 | ||
| XM_005551364.2 | XP_005551421.1 | 5 - 6 | 864 | 112 | ||
| XM_015448377.1 | XP_015303863.1 | 6 - 7 | 1076 | 112 | ||
| XM_015448378.1 | XP_015303864.1 | 6 - 7 | 875 | 112 | ||
| GenBank mRNA | AB168365.1 | - | 5 - 6 | 906 | 112 | |
| AB178959.1 | - | 5 - 6 | 908 | 112 | ||
More Information
Set Membership: |
|

TBP
Genomic Map