Assay Details
| Gene Symbol: WNT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542309.2 | XP_005542366.1 | 3 - 4 | 1220 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WNT2B| Gene Symbol: WNT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542309.2 | XP_005542366.1 | 3 - 4 | 1220 | 76 | |
Set Membership: |
|