Assay Details
| Gene Symbol: PCBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547290.1 | XP_005547347.1 | 5 - 6 | 516 | 59 | |
| XM_005547292.2 | XP_005547349.1 | 5 - 6 | 482 | 59 | ||
| XM_005547293.2 | XP_005547350.1 | 4 - 5 | 334 | 59 | ||
| XM_005547295.2 | XP_005547352.1 | 4 - 5 | 374 | 59 | ||
| XM_005547296.2 | XP_005547353.1 | 4 - 5 | 1601 | 59 | ||
| XM_015445968.1 | XP_015301454.1 | 5 - 6 | 389 | 59 | ||
| XM_015445969.1 | XP_015301455.1 | 5 - 6 | 430 | 59 | ||
| XM_015445971.1 | XP_015301457.1 | 4 - 5 | 1063 | 59 | ||
| XM_015445972.1 | XP_015301458.1 | 5 - 6 | 355 | 59 | ||
| XM_015445973.1 | XP_015301459.1 | 3 - 4 | 201 | 59 | ||
More Information
Set Membership: |
|

PCBP4
Genomic Map