Assay Details
| Gene Symbol: C8H8orf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563491.2 | XP_005563548.1 | 9 - 10 | 1669 | 103 | |
| XM_015454933.1 | XP_015310419.1 | 9 - 10 | 1669 | 103 | ||
| XM_015454934.1 | XP_015310420.1 | 9 - 10 | 1669 | 103 | ||
| XM_015454935.1 | XP_015310421.1 | 8 - 9 | 1541 | 103 | ||
More Information
Set Membership: |
|

C8H8orf34
Genomic Map