Assay Details
| Gene Symbol: TIMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287306.1 | NP_001274235.1 | 1 - 2 | 150 | 57 | |
| GenBank mRNA | AY207385.1 | - | 1 - 2 | 150 | 57 | |
More Information
Set Membership: |
|

TIMP3
Genomic Map