Assay Details
| Gene Symbol: FBXO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283335.1 | NP_001270264.1 | 5 - 6 | 683 | 78 | |
| XM_005578238.2 | XP_005578295.1 | 5 - 6 | 722 | 78 | ||
| XM_015434130.1 | XP_015289616.1 | 5 - 6 | 722 | 78 | ||
| GenBank mRNA | AB172188.1 | - | 5 - 6 | 683 | 78 | |
More Information
Set Membership: |
|

FBXO3
Genomic Map