Assay Details
| Gene Symbol: ME3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283412.1 | NP_001270341.1 | 12 - 13 | 1808 | 63 | |
| GenBank mRNA | AB170878.1 | - | 12 - 13 | 1808 | 63 | |
More Information
Set Membership: |
|

ME3
Genomic Map