Assay Details
| Gene Symbol: IDH3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287636.1 | NP_001274565.1 | 2 - 3 | 139 | 68 | |
| XM_005568512.1 | XP_005568569.1 | 3 - 4 | 547 | 68 | ||
| XM_005568513.1 | XP_005568570.1 | 3 - 4 | 547 | 68 | ||
| XM_005568514.2 | XP_005568571.1 | 3 - 4 | 547 | 68 | ||
| GenBank mRNA | AB169839.1 | - | 2 - 3 | 146 | 68 | |
| AB170260.1 | - | 2 - 3 | 139 | 68 | ||
| X82632.1 | - | 1 | 113 | 68 | ||
More Information
Set Membership: |
|

IDH3B
Genomic Map