Assay Details
| Gene Symbol: CAPZA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542336.2 | XP_005542393.1 | 6 - 7 | 1182 | 67 | |
| XM_005542337.2 | XP_005542394.1 | 6 - 7 | 503 | 67 | ||
| GenBank mRNA | AB168237.1 | - | 6 - 7 | 545 | 67 | |
More Information
Set Membership: |
|

CAPZA1
Genomic Map