Assay Details
Gene Symbol: PCDH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005554640.2 | XP_005554697.1 | 1 - 2 | 3176 | 76 | |
XM_005554641.2 | XP_005554698.1 | 1 - 2 | 3176 | 76 | ||
GenBank mRNA | AB220501.1 | - | 1 - 2 | 1079 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
