Assay Details
Gene Symbol: GPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287712.1 | NP_001274641.1 | 4 - 5 | 1061 | 61 | |
GenBank mRNA | AB169665.1 | - | 4 - 5 | 1061 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
