Assay Details
| Gene Symbol: BUB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566671.1 | XP_005566728.1 | 7 - 8 | 1212 | 75 | |
| GenBank mRNA | AB174420.1 | - | 7 - 8 | 1059 | 75 | |
More Information
Set Membership: |
|

BUB3
Genomic Map