Assay Details
| Gene Symbol: BTN2A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553788.2 | XP_005553845.1 | 5 - 6 | 1045 | 122 | |
| XM_005553789.2 | XP_005553846.1 | 5 - 6 | 1154 | 122 | ||
| XM_015449774.1 | XP_015305260.1 | 5 - 6 | 1083 | 122 | ||
| XM_015449775.1 | XP_015305261.1 | 4 - 5 | 988 | 122 | ||
| XM_015449776.1 | XP_015305262.1 | 5 - 6 | 1156 | 122 | ||
| XM_015449777.1 | XP_015305263.1 | 5 - 6 | 1086 | 122 | ||
| XM_015449778.1 | XP_015305264.1 | 5 - 6 | 1048 | 122 | ||
| XM_015449779.1 | XP_015305265.1 | 5 - 6 | 1154 | 122 | ||
| XM_015449780.1 | XP_015305266.1 | 5 - 6 | 1153 | 122 | ||
| XM_015449781.1 | XP_015305267.1 | 5 - 6 | 1153 | 122 | ||
More Information
Set Membership: |
|

BTN2A2
Genomic Map