Assay Details
| Gene Symbol: AGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550049.1 | XP_005550106.1 | 4 - 5 | 435 | 111 | |
| XM_005550050.2 | XP_005550107.1 | 4 - 5 | 405 | 111 | ||
More Information
Set Membership: |
|

AGR2
Genomic Map