Assay Details
| Gene Symbol: PID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319552.1 | NP_001306481.1 | 3 - 4 | 374 | 61 | |
| XM_005574519.2 | XP_005574576.1 | 2 - 3 | 775 | 61 | ||
| XM_015432914.1 | XP_015288400.1 | 2 - 3 | 442 | 61 | ||
| GenBank mRNA | AB171972.1 | - | 3 - 4 | 374 | 61 | |
More Information
Set Membership: |
|

PID1
Genomic Map