Assay Details
| Gene Symbol: PPP2R2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283453.1 | NP_001270382.1 | 6 - 7 | 936 | 78 | |
| XM_005562881.2 | XP_005562938.1 | 6 - 7 | 919 | 78 | ||
| XM_015454361.1 | XP_015309847.1 | 7 - 8 | 993 | 78 | ||
| GenBank mRNA | AB168667.1 | - | 6 - 7 | 936 | 78 | |
More Information
Set Membership: |
|

PPP2R2A
Genomic Map