Assay Details
| Gene Symbol: MAPK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559576.2 | XP_005559633.1 | 3 - 4 | 1517 | 130 | |
| XM_005559577.2 | XP_005559634.1 | 3 - 4 | 1741 | 130 | ||
| XM_015452836.1 | XP_015308322.1 | 3 - 4 | 1737 | 130 | ||
| XM_015452837.1 | XP_015308323.1 | 3 - 4 | 1540 | 130 | ||
| GenBank mRNA | AB168593.1 | - | 1 - 2 | 161 | 130 | |
More Information
Set Membership: |
|

MAPK6
Genomic Map