Assay Details
| Gene Symbol: PQBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593501.2 | XP_005593558.1 | 4 - 5 | 491 | 58 | |
| XM_005593502.2 | XP_005593559.1 | 4 - 5 | 495 | 58 | ||
| XM_005593504.2 | XP_005593561.1 | 4 - 5 | 918 | 58 | ||
| XM_005593505.2 | XP_005593562.1 | 4 - 5 | 448 | 58 | ||
| XM_005593506.2 | XP_005593563.1 | 4 - 5 | 400 | 58 | ||
| XM_005593507.2 | XP_005593564.1 | 4 - 5 | 404 | 58 | ||
More Information
Set Membership: |
|

PQBP1
Genomic Map