Assay Details
| Gene Symbol: CCNQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594905.2 | XP_005594962.1 | 1 - 2 | 223 | 69 | |
| XM_005594906.2 | XP_005594963.1 | 1 - 2 | 223 | 69 | ||
More Information
Set Membership: |
|

CCNQ
Genomic Map