Assay Details
| Gene Symbol: HDHD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284981.1 | NP_001271910.1 | 6 - 7 | 766 | 60 | |
| XM_015456926.1 | XP_015312412.1 | 6 - 7 | 642 | 60 | ||
| GenBank mRNA | AB174689.1 | - | 6 - 7 | 766 | 60 | |
More Information
Set Membership: |
|

HDHD5
Genomic Map