Assay Details
| Gene Symbol: ZFAND2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574345.2 | XP_005574402.1 | 7 - 8 | 664 | 57 | |
| XM_005574346.2 | XP_005574403.1 | 7 - 8 | 662 | 57 | ||
| XM_005574347.2 | XP_005574404.1 | 6 - 7 | 715 | 57 | ||
| XM_015432844.1 | XP_015288330.1 | 6 - 7 | 691 | 57 | ||
| XM_015432845.1 | XP_015288331.1 | 7 - 8 | 653 | 57 | ||
| XM_015432846.1 | XP_015288332.1 | 6 - 7 | 607 | 57 | ||
More Information
Set Membership: |
|

ZFAND2B
Genomic Map