Assay Details
| Gene Symbol: ALAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285030.1 | NP_001271959.1 | 6 - 7 | 1132 | 65 | |
| XM_005547319.2 | XP_005547376.1 | 6 - 7 | 1191 | 65 | ||
| XM_015445337.1 | XP_015300823.1 | 5 - 6 | 907 | 65 | ||
| GenBank mRNA | AB169255.1 | - | 6 - 7 | 1132 | 65 | |
More Information
Set Membership: |
|

ALAS1
Genomic Map