Assay Details
| Gene Symbol: NDE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283612.1 | NP_001270541.1 | 2 - 3 | 300 | 65 | |
| GenBank mRNA | AB172487.1 | - | 2 - 3 | 300 | 65 | |
More Information
Set Membership: |
|

NDE1
Genomic Map