Assay Details
| Gene Symbol: SLC33A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284569.1 | NP_001271498.1 | 1270 | 82 | ||
| GenBank mRNA | AB172193.1 | - | 1270 | 82 | ||
| AB172658.1 | - | 1400 | 82 | |||
More Information
Set Membership: |
|

SLC33A1
Genomic Map