Assay Details
| Gene Symbol: SERBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543075.2 | XP_005543132.1 | 1 - 2 | 780 | 104 | |
| XM_005543076.2 | XP_005543133.1 | 1 - 2 | 780 | 104 | ||
| XM_005543077.2 | XP_005543134.1 | 1 - 2 | 780 | 104 | ||
| XM_005543078.2 | XP_005543135.1 | 1 - 2 | 780 | 104 | ||
| GenBank mRNA | AB168184.1 | - | 1 - 2 | 436 | 104 | |
| AB168186.1 | - | 1 - 2 | 436 | 104 | ||
More Information
Set Membership: |
|

SERBP1
Genomic Map