Assay Details
| Gene Symbol: NKX2-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561100.2 | XP_005561157.1 | 1 - 2 | 350 | 123 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005561100.2 transcripts.
Set Membership: |
|

NKX2-1
Genomic Map