Assay Details
| Gene Symbol: NOP56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284554.1 | NP_001271483.1 | 5 - 6 | 1084 | 73 | |
| XM_005568518.2 | XP_005568575.1 | 5 - 6 | 658 | 73 | ||
| GenBank mRNA | AB066543.1 | - | 5 - 6 | 1084 | 73 | |
More Information
Set Membership: |
|

NOP56
Genomic Map