Assay Details
| Gene Symbol: ADAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283444.1 | NP_001270373.1 | 8 - 9 | 1692 | 66 | |
| XM_015442707.1 | XP_015298193.1 | 7 - 8 | 2497 | 66 | ||
| XM_015442708.1 | XP_015298194.1 | 7 - 8 | 1255 | 66 | ||
| GenBank mRNA | AB168782.1 | - | 8 - 9 | 1692 | 66 | |
More Information
Set Membership: |
|

ADAT1
Genomic Map