Assay Details
| Gene Symbol: LRRC43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319518.1 | NP_001306447.1 | 1 - 2 | 154 | 71 | |
| XM_015431675.1 | XP_015287161.1 | 1 - 2 | 169 | 71 | ||
| XM_015431676.1 | XP_015287162.1 | 1 - 2 | 171 | 71 | ||
| GenBank mRNA | AB070096.1 | - | 1 - 2 | 154 | 71 | |
More Information
Set Membership: |
|

LRRC43
Genomic Map